missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TIMP2 / TIMP-2 Protein
A DNA sequence encoding the Homo sapiens (Human) TIMP2 / TIMP-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8571-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003246.1 | |
TIMP2 / TIMP-2 Protein | |
Research Use Only | |
TIMP2 | |
Recombinant Protein | |
E. coli | |
SPVHPQQAFC NADVVIRAKA VSEKEVDSGN DIYGNPIKRI QYEIKQIKMF KGPEKDIEFI YTAPSSAVCG VSLDVGGKKE YLIAGKAEGD GKMHITLCDF IVPWDTLSTT QKKSLNHRYQ MGCECKITRC PMIPCYISSP DECLWMDWVT EKNINGHQAK FFACIKRSDG SCAWYRGAAP PKQEFLDIED P | |
Greater than 90% as determined by SDS-PAGE. |
7077 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |