missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TIMP2 / TIMP-2 Protein

A DNA sequence encoding the Homo sapiens (Human) TIMP2 / TIMP-2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8571-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16035351

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_003246.1
TIMP2 / TIMP-2 Protein
Research Use Only
TIMP2
Recombinant Protein
E. coli
SPVHPQQAFC NADVVIRAKA VSEKEVDSGN DIYGNPIKRI QYEIKQIKMF KGPEKDIEFI YTAPSSAVCG VSLDVGGKKE YLIAGKAEGD GKMHITLCDF IVPWDTLSTT QKKSLNHRYQ MGCECKITRC PMIPCYISSP DECLWMDWVT EKNINGHQAK FFACIKRSDG SCAWYRGAAP PKQEFLDIED P
Greater than 90% as determined by SDS-PAGE.
7077
50 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers