missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TIE1 Fc Protein
A cDNA sequence encoding the TIE1 Fc was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13750-1mg
Additional Details : Weight : 0.01000kg
Specifications
TIE1 Fc Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MVWRVPPFLL PILFLASHVG AAVDLTLLAN LRLTDPQRFF LTCVSGEAGA GRGSDAWGPP LLLEKDDRIV RTPPGPPLRL ARNGSHQVTL RGFSKPSDLV GVFSCVGGAG ARRTRVIYVH NSPGAHLLPD KVTHTVNKGD TAVLSARVHK EKQTDVIWKS NGSYFYTLDW HEAQDGRFLL QLPNVQPPSS GIYSATYLEA SPLGSAFFRL IVRGCGAGRW GPGCTKECPG CLHGGVCHDH DGECVCPPGF TGTRCEQACR EGRFGQSCQE QCPGISGCRG LTFCLPDPYG CSCGSGWRGS QCQEACAPGH FGADCRLQCQ CQNGGTCDRF SGCVCPSGWH GVHCEKSDRI PQILNMASEL EFNLETMPRI NCAAAGNPFP VRGSIELRKP DGTVLLSTKA IVEPEKTTAE FEVPRLVLAD SGFWECRVST SGGQDSRRFK VNVKVPPVPL AAPRLLTKQS RQLVVSPLVS FSGDGPISTV RLHYRPQDST MDWSTIVVDP SENVTLMNLR PKTGYSVRVQ LSRPGEGGEG AWGPPTLMTT DCPEPLLQPW LEGWHVEGTD RLRVSWSLPL VPGPLVGDGF LLRLWDGTRG QERRENVSSP QARTALLTGL TPGTHYQLDV QLYHCTLLGP ASPPAHVLLP PSGPPAPRHL HAQALSDSEI QLTWKHPEAL PGPISKYVVE VQVAGGAGDP LWIDVDRPEE TSTIIRGLNA STRYLFRMRA SIQGLGDWSN TVEESTLGNG LQAEGPVQES RAAEEGLDQQ LILAVVGSVS ATCLTILAAL LTLVCIRRSC LHRRRTFTYQ SGSGEETILQ FSSGTLTLTR RPKLQPEPLS YPVLEWEDIT FEDLIGEGNF GQVIRAMIKK DGLKMNAAIK MLKEYASEND HRDFAGELEV LCKLGHHPNI INLLGACKNR | |
Greater than 90.0% as determined by:(A) Analysis by RP-HPLC. (B) Analysis by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
TIE-1 Fc Chimera was lyophilized from a concentrated (1 mg/ml) sterile solution containing 20mM Tris, 0.5M NaCl, 10% Sucrose. |