missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human THYN1 Protein
A cDNA sequence encoding the THYN1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13747-20ug
Additional Details : Weight : 0.01000kg
Specifications
THYN1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSRPRKRLAG TSGSDKGLSG KRTKTENSGE ALAKVEDSNP QKTSATKNCL KNLSSHWLMK SEPESRLEKG VDVKFSIEDL KAQPKQTTCW DGVRNYQARN FLRAMKLGEE AFFYHSNCKE PGIAGLMKIV KEAYPDHTQF EKNNPHYDPS SKEDNPKWSM VDVQFVRMMK RFIPLAELKS YHQAHKATGG PLKNMVLFTR QRLSIQPLTQ EEFDFVLSLE EKEPS | |
Greater than 95% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The THYN1 protein solution (0.5 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0) 1mM DTT, 100mM NaCl and 20% glycerol. |