missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Thrombopoietin / THPO Protein
Recombinant Protein
£80.80 - £4515.00
Specifications
Accession Number | NP_000451.1 |
---|---|
Gene ID (Entrez) | 7066 |
Name | Thrombopoietin / THPO Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15955739
|
enQuireBio™
QP5227-2UG |
2 μg |
£80.80
2µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935739
|
enQuireBio™
QP5227-10UG |
10 μg |
£191.00
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965739
|
enQuireBio™
QP5227-500UG |
500 μg |
£2755.00
500µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945739
|
enQuireBio™
QP5227-1MG |
1 mg |
£4515.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_000451.1 | |
Thrombopoietin / THPO Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 5 ng/mL in the MO7e cell proliferaiton assay. The bioactivity acceptance criteria is set at 2 x 10^5 units/mg. | |
Human | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
7066 | |
Research Use Only | |
THPO | |
Recombinant Protein | |
E. coli | |
MSPAPPACDL RVLSKLLRDS HVLHSRLSQC PEVHPLPTPV LLPAVDFSLG EWKTQMEETK AQDILGAVTL LLEGVMAARG QLGPTCLSSL LGQLSGQVRL LLGALQSLLG TQLPPQGRTT AHKDPNAIFL SFQHLLRGKV RFLMLVGGST LCVRRAPPTT AVPSRTSLVL TLNEL | |
Greater than 90 as determined by SDS-PAGE. |