missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Thioredoxin / TXN / SASP Protein

A DNA sequence encoding the Homo sapiens (Human) Thioredoxin / TXN / SASP Protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8427-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16004561

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001231867.1
Thioredoxin / TXN / SASP Protein
Research Use Only
TXN
Recombinant Protein
E. coli
VKQIESKTAF QEALDAAGDK LVVVDFSATW CGPCKMIKPF FHSLSEKYSN VIFLEVDVDD CQDVASECEV KCMPTFQFFK KGQKVGEFSG ANKEKLEATI NELV
Greater than 90% as determined by SDS-PAGE.
7295
500 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers