missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human THAP3 Protein

A cDNA sequence encoding the THAP3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13740-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15964549

  • £99.15 / 2µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

THAP3 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMPKSCAA RQCCNRYSSR RKQLTFHRFP FSRPELLKEW VLNIGRGNFK PKQHTVICSE HFRPECFSAF GNRKNLKHNA VPTVFAFQDP TQQVRENTDP ASERGNASSS QKEKVLPEAG AGEDSPGRNM DTALEELQLP PNAEGHVKQV SPRRPQATEA VGRPTGPAGL RRTPNKQPSD HSYALLDLDS LKKKLFLTLK ENEKLRKRLQ AQRLVMRRMS SRLRACKGHQ GLQARLGPEQ QS
Greater than 85% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The THAP3 solution contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 2mM DTT and 50% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers