missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TGFBR2 Protein
A cDNA sequence encoding the TGFBR2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13734-10ug
Additional Details : Weight : 0.01000kg
Specifications
TGFBR2 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
TIPPHVQKSV NNDMIVTDNN GAVKFPQLCK FCDVRFSTCD NQKSCMSNCS ITSICEKPQE VCVAVWRKND ENITLETVCH DPKLPYHDFI LEDAASPKCI MKEKKKPGET FFMCSCSSDE CNDNIIFSEE YNTSNPDLLL VIFQLEPKSC DKTHTCPPCP APELLGGPSV FLFPPKPKDT LMISRTPEVT CVVVDVSHED PEVKFNWYVD GVEVHNAKTK PREEQYNSTY RVVSVLTVLH QDWLNGKEYK CKVSNKALPA PIEKTISKAK GQPREPQVYT LPPSRDELTK NQVSLTCLVK GFYPSDIAVE WESNGQPENN YKTTPPVLDS DGSFFLYSKL TVDKSRWQQG NVFSCSVMHE ALHNHYTQKS LSLSPGKHHH HHH | |
Greater than 95% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The TGFBR2 solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |