missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TGFBI (135 Amino Acids Protein
A cDNA sequence encoding the TGFBI (135 Amino Acids was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13733-EC-1mg
Additional Details : Weight : 0.01000kg
Specifications
TGFBI (135 Amino Acids Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGTVMDVLKG DNRFSMLVAA IQSAGLTETL NREGVYTVFA PTNEAFRALP PRERSRLLGD AKELANILKY HIGDEILVSG GIGALVRLKS LQGDKLEVSL KNNVVSVNKE PVAEPDIMAT NGVVHVITNV LQPPA | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The TGFBI recombinant Human is formulated 20mM Tris-HCl pH 8. |