missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TEF Protein
A cDNA sequence encoding the TEF was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13717-10ug
Additional Details : Weight : 0.01000kg
Specifications
7008 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSDAGGG KKPPVDPQAG PGPGPGRAAG ERGLSGSFPL VLKKLMENPP REARLDKEKG KEKLEEDEAA AASTMAVSAS LMPPIWDKTI PYDGESFHLE YMDLDEFLLE NGIPASPTHL AHNLLLPVAE LEGKESASSS TASPPSSSTA IFQPSETVSS TESSLEKERE TPSPIDPNCV EVDVNFNPDP ADLVLSSVPG GELFNPRKHK FAEEDLKPQP MIKKAKKVFV PDEQKDEKYW TRRKKNNVAA KRSRDARRLK ENQITIRAAF LEKENTALRT EVAELRKEVG KCKTIVSKYE TKYGPL | |
Greater than 85% as determined by SDS-PAGE. |
TEF Protein | |
Research Use Only | |
TEF | |
Human | |
His | |
The TEF solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |