missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TCEAL8 Protein
A cDNA sequence encoding the TCEAL8 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13709-1mg
Additional Details : Weight : 0.01000kg
Specifications
TCEAL8 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MQKSCEENEG KPQNMPKAEE DRPLEDVPQE AEGNPQPSEE GVSQEAEGNP RGGPNQPGQG FKEDTPVRHL DPEEMIRGVD ELERLREEIR RVRNKFVMMH WKQRHSRSRP YPVCFRPLEH HHHHH | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TCEAL8 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer, pH 8.0, 20% glycerol, 1mM DTT and 200mM NaCl. |