missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TBCB Protein
A cDNA sequence encoding the TBCB was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13696-25ug
Additional Details : Weight : 0.01000kg
Specifications
TBCB Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMEVTGV SAPTVTVFIS SSLNTFRSEK RYSRSLTIAE FKCKLELLVG SPASCMELEL YGVDDKFYSK LDQEDALLGS YPVDDGCRIH VIDHSGARLG EYEDVSRVEK YTISQEAYDQ RQDTVRSFLK RSKLGRYNEE ERAQQEAEAA QRLAEEKAQA SSIPVGSRCE VRAAGQSPRR GTVMYVGLTD FKPGYWIGVR YDEPLGKNDG SVNGKRYFEC QAKYGAFVKP AVVTVGDFPE EDYGLDEI | |
Greater than 90% as determined by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The TBCB solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM Nacl and 20% glycerol. |