missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TBCA Protein
A cDNA sequence encoding the TBCA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13695-5ug
Additional Details : Weight : 0.01000kg
Specifications
TBCA Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MADPRVRQIK IKTGVVKRLV KEKVMYEKEA KQQEEKIEKM RAEDGENYDI KKQAEILQES RMMIPDCQRR LEAAYLDLQR ILENEKDLEE AEEYKEARLV LDSVKLEA | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The TBCA solution contains 20mM Tris-HCl buffer pH 7.5, 1mM DTT and 10% glycerol. |