missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TARS Protein
A cDNA sequence encoding the TARS was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13689-10ug
Additional Details : Weight : 0.01000kg
Specifications
TARS Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MFEEKASSPS GKMGGEEKPI GAGEEKQKEG GKKKNKEGSG DGGRAELNPW PEYIYTRLEM YNILKAEHDS ILAEKAEKDS KPIKVTLPDG KQVDAESWKT TPYQIACGIS QGLADNTVIA KVNNVVWDLD RPLEEDCTLE LLKFEDEEAQ AVYWHSSAHI MGEAMERVYG GCLCYGPPIE NGFYYDMYLE EGGVSSNDFS SLEALCKKII KEKQAFERLE VKKETLLAMF KYNKFKCRIL NEKVNTPTTT VYRCGPLIDL CRGPHVRHTG KIKALKIHKN SSTYWEGKAD METLQRIYGI SFPDPKMLKE WEKFQEEAKN RDHRKIGRDQ ELYFFHELSPGSCFFLPKGA YIYNALIEFI RSEYRKRGFQ EVVTPNIFNS RLWMTSGHWQ HYSENMFSFE VEKELFALKP MNCPGHCLMF DHRPRSWREL PLRLADFGVL HRNELSGALT GLTRVRRFQQ DDAHIFCAME QIEDEIKGCL DFLRTVYSVF GFSFKLNLST RPEKFLGDIE VWDQAEKQLE NSLNEFGEKW ELNSGDGAFY GPKIDIQIKD AIGRYHQCAT IQLDFQLPIR FNLTYVSHDG DDKKRPVIVH RAILGSVERM IAILTENYGG KWPFWLSPRQ VMVVPVGPTC DEYAQKVRQQ FHDAKFMADI DLDPGCTLNK KIRNAQLAQY NFILVVGEKE KISGTVNIRT RDNKVHGERT ISETIERLQQ LKEFRSKQAE EEF | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The TARS solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 20% glycerol and 150mM NaCl. |