missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TAF9 Protein
A cDNA sequence encoding the TAF9 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | TAF9 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15984379
|
enQuireBio™
QP13686-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15974379
|
enQuireBio™
QP13686-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964379
|
enQuireBio™
QP13686-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
TAF9 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TAF9 Human solution (0.5 mg/1ml) containing 20mM Trsi HCL pH 8.0, 1mM DTT and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MLLPNILLTG TPGVGKTTLG KELASKSGLK YINVGDLARE EQLYDGYDEE YDCPILDEDR VVDELDNQMR EGGVIVDYHG CDFFPERWFH IVFVLRTDTN VLYERLETRG YNEKKLTDNI QCEIFQVLYE EATASYKEEI VHQLPSNKPE ELENNVDQIL KWIEQWIKDH NS | |
Greater than 90% as determined by SDS-PAGE. |