missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TAF15 Protein
A cDNA sequence encoding the TAF15 was constructed and used to recombinantly synthesize the protein.
£99.15 - £3965.00
Specifications
Name | TAF15 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15954379
|
enQuireBio™
QP13685-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944379
|
enQuireBio™
QP13685-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15934379
|
enQuireBio™
QP13685-1MG |
1 mg |
£3965.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
TAF15 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The TAF15 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 30% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSSYHSQRE NYSHHTQDDR RDVSRYGEDN RGYGGSQGGG RGRGGYDKDG RGPMTGSSGG DRGGFKNFGG HRDYGPRTDA DSESDNSDNN TIFVQGLGEG VSTDQVGEFF KQIGIIKTNK KTGKPMINLY TDKDTGKPKG EATVSFDDPP SAKAAIDWFD GKEFHGNIIK VSFATRRPEF MRGGGSGGGR RGRGGYRGRG GFQGRGGDPK SGDWVCPNPS CGNMNFARRN SCNQCNEPRP EDSRPSGGDF RGRGYGGERG YR | |
Greater than 90% as determined by SDS-PAGE. |