missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TACSTD2 Protein

A cDNA sequence encoding the TACSTD2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13683-HIS-25ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15954369

  • £151.37 / 25µg
Estimated Shipment: 12-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TACSTD2 Protein
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
TACSTD2 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSQDNCTCP TNKMTVCSPD GPGGRCQCRA LGSGMAVDCS TLTSKCLLLK ARMSAPKNAR TLVRPSEHAL VDNDGLYDPD CDPEGRFKAR QCNQTSVCWC VNSVGVRRTD KGDLSLRCDE LVRTHHILID LRHRPTAGAF NHSDLDAELR RLFRERYRLH PKFVAAVHYE QPTIQIELRQ NTSQKAAGDV DIGDAAYYFE RDIKGESLFQ GRGGLDLRVR GEPLQVERTL IYYLDEIPPK FSMKRLT
Videos
SDS
Documents
Certificates
Special Offers

Special Offers