missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TAC3 Protein

A cDNA sequence encoding the TAC3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13680-25ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15964359

  • £151.37 / 25µg
Estimated Shipment: 13-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TAC3 Protein
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The TAC3 solution contains 20mM Tris-HCl buffer (pH 8.0) and 20% glycerol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH QSFGAVCKEP QEEVVPGGGR SKRDPDLYQL LQRLFKSHSS LEGLLKALSQ ASTDPKESTS PEKRDMHDFF VGLMGKRSVQ PDSPTDVNQE NVPSFGILKY PPRAE
Videos
SDS
Documents
Certificates
Special Offers

Special Offers