missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SYK Protein

A cDNA sequence encoding the SYK was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13656-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15954289

  • £99.15 / 2µg
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

6850
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Human (HEK293)
MHHHHHHDYKDDDDKKLMASSGMADSANHLPFFFGNITREEAEDYLVQGGMSDGL YLLRQSRNYLGGFALSVAHGRKAHHYTIERELNGTYAIAGGRTHASPADLCHYHSQES DGLVCLLKKPFNRPQGVQPKTGPFEDLKENLIREYVKQTWNLQGQALEQAIISQKPQL EKLIATTAHEKMPWFHGKISREESEQIVLIGSKTNGKFLIRARDNNGSYALCLLHEGKV LHYRIDKDKTGKLSIPEGKKFDTLWQLVEHYSYKADGLLRVLTVPCQKIGTQGNVNFG GRPQLPGSHPATWSAGGIISRIKSYSFPKPGHRKSSPAQGNRQESTVSFNPYEPELA PWAADKGPQREALPMDTEVYESPYADPEEIRPKEVYLDRKLLTLEDKELGSGNFGTV KKGYYQMKKVVKTVAVKILKNEANDPALKDELLAEANVMQQLDNPYIVRMIGICEAES WMLVMEMAELGPLNKYLQQNRHVKDKNIIELVHQVSMGMKYLEESNFVHRDLAARN VLLVTQHYAKISDFGLSKALRADENYYKAQTHGKWPVKWYAPECINYYKFSSKSDVW SFGVLMWEAFSYGQKPYRGMKGSEVTAMLEKGERMGCPAGCPREMYDLMNLCWT YDVENRPGFAAVELRLRNYYYDVVN
Greater than 85.0% as determined by SDS-PAGE.
SYK Protein
Research Use Only
SYK
Human
Flag Tag
SYK protein is supplied in 50mM Tris pH 7.5, 300mM NaCl and 10% Glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers