missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SYK Protein
A cDNA sequence encoding the SYK was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13656-10ug
Additional Details : Weight : 0.01000kg
Specifications
6850 | |
Greater than 85.0% as determined by SDS-PAGE. | |
Research Use Only | |
SYK | |
Human | |
Flag Tag | |
SYK protein is supplied in 50mM Tris pH 7.5, 300mM NaCl and 10% Glycerol. |
SYK Protein | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
HEK 293 | |
MHHHHHHDYKDDDDKKLMASSGMADSANHLPFFFGNITREEAEDYLVQGGMSDGL YLLRQSRNYLGGFALSVAHGRKAHHYTIERELNGTYAIAGGRTHASPADLCHYHSQES DGLVCLLKKPFNRPQGVQPKTGPFEDLKENLIREYVKQTWNLQGQALEQAIISQKPQL EKLIATTAHEKMPWFHGKISREESEQIVLIGSKTNGKFLIRARDNNGSYALCLLHEGKV LHYRIDKDKTGKLSIPEGKKFDTLWQLVEHYSYKADGLLRVLTVPCQKIGTQGNVNFG GRPQLPGSHPATWSAGGIISRIKSYSFPKPGHRKSSPAQGNRQESTVSFNPYEPELA PWAADKGPQREALPMDTEVYESPYADPEEIRPKEVYLDRKLLTLEDKELGSGNFGTV KKGYYQMKKVVKTVAVKILKNEANDPALKDELLAEANVMQQLDNPYIVRMIGICEAES WMLVMEMAELGPLNKYLQQNRHVKDKNIIELVHQVSMGMKYLEESNFVHRDLAARN VLLVTQHYAKISDFGLSKALRADENYYKAQTHGKWPVKWYAPECINYYKFSSKSDVW SFGVLMWEAFSYGQKPYRGMKGSEVTAMLEKGERMGCPAGCPREMYDLMNLCWT YDVENRPGFAAVELRLRNYYYDVVN |