missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Surfactant-associated protein 1

A DNA sequence encoding the Homo sapiens (Human) Surfactant-associated protein 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6680-ma-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001094811.1
Surfactant-associated protein 1
Research Use Only
SFTA3
Recombinant Protein
Mammal
MRAGFSDFQL IRDQVLFLQD QAQRLTEWLQ LSGFENPVSE STTLCLRERE KRIPTCVAVC VPSPGTVHTA LLHPTTLSQS RSSSEAKMLI IHTA
Greater than 90% as determined by SDS-PAGE.
253970
50 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers