missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SUMO3 Protein
A cDNA sequence encoding the SUMO3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13650-20ug
Additional Details : Weight : 0.01000kg
Specifications
SUMO3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSEEKPKEGV KTENDHINLK VAGQDGSVVQ FKIKRHTPLS KLMKAYCERQ GLSMRQIRFR FDGQPINETD TPAQLEMEDE DTIDVFQQQT GG | |
Greater than 95% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SUMO3 protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0) 0.1M NaCl,1mM DTT and 20% glycerol. |