missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SUMO-conjugating enzyme UBC9 Protein
A DNA sequence encoding the Homo sapiens (Human) SUMO-conjugating enzyme UBC9, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8322-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003336.1 | |
SUMO-conjugating enzyme UBC9 Protein | |
Research Use Only | |
UBE2I | |
Recombinant Protein | |
E. coli | |
MSGIALSRLA QERKAWRKDH PFGFVAVPTK NPDGTMNLMN WECAIPGKKG TPWEGGLFKL RMLFKDDYPS SPPKCKFEPP LFHPNVYPSG TVCLSILEED KDWRPAITIK QILLGIQELL NEPNIQDPAQ AEAYTIYCQN RVEYEKRVRA QAKKFAP | |
Greater than 90% as determined by SDS-PAGE. |
7329 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |