missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STUB1 Protein
A cDNA sequence encoding the STUB1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13629-1mg
Additional Details : Weight : 0.01000kg
Especificaciones
STUB1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKGKEEKEGG ARLGAGGGSP EKSPSAQELK EQGNRLFVGR KYPEAAACYG RAITRNPLVA VYYTNRALCY LKMQQHEQAL ADCRRALELD GQSVKAHFFL GQCQLEMESY DEAIANLQRA YSLAKEQRLN FGDDIPSALR IAKKKRWNSI EERRIHQESE LHSYLSRLIA AERERELEEC QRNHEGDEDD SHVRAQQACI EAKHDKYMAD MDELFSQVDE KRKKRDIPDY LCGKISFELM REPCITPSGI TYDRKDIEEH LQRVGHFDPV TRSPLTQEQL IPNLAMKEVI DAFISENGWV EDY | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The STUB1 protein solution contains 20mM Tris-HCl, pH 7.5, 10% glycerol and 5mM DTT. |