missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human STEAP4 Protein

A cDNA sequence encoding the STEAP4 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13614-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15944149

  • £2975.00 / 1mg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

STEAP4 Protein
Research Use Only
Recombinant Protein
E. coli
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMEKT CIDALPLTMN SSEKQETVCI FGTGDFGRSL GLKMLQCGYS VVFGSRNPQK TTLLPSGAEV LSYSEAAKKS GIIIIAIHRE HYDFLTELTE VLNGKILVDI SNNLKINQYP ESNAEYLAHL VPGAHVVKAF NTISAWALQS GALDASRQ
Greater than 90.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
STEAP4 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers