missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STBD1 Protein
A cDNA sequence encoding the STBD1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £3965.00
Specifications
Name | STBD1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15974139
|
enQuireBio™
QP13612-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964139
|
enQuireBio™
QP13612-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954139
|
enQuireBio™
QP13612-1MG |
1 mg |
£3965.00
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
STBD1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The STBD1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSRGGPGDT GKDGDAEQEK DAPLGGAAIP GGHQSGSSGL SPGPSGQELV TKPEHLQESN GHLISKTKDL GKLQAASWRL QNPSREVCDN SREHVPSGQF PDTEAPATSE TSNSRSYSEV SRNESLESPM GEWGFQKGQE ISAKAATCFA EKLPSSNLLK NRAKEEMSLS DLNSQDRVDH EEWEMVPRHS SWGDVGVGGS LKAPVLNLNQ GMDNGRSTLV EARGQQVHGK MERVAVMPAG SQQVSVRFQV HYVTSTDVQF IAVTGDHECL GRWNTYIPLH YNKDGFWSHS IFLPADTVVE WKFVLVENGG VTRWEECSNR FLETGHEDKV VHAWWGIH | |
Greater than 90% as determined by SDS-PAGE. |