missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STAR Protein
A cDNA sequence encoding the STAR was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13609-1mg
Additional Details : Weight : 0.01000kg
Specifications
STAR Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MEETLYSDQE LAYLQQGEEA MQKALGILSN QEGWKKESQQ DNGDKVMSKV VPDVGKVFRL EVVVDQPMER LYEELVERME AMGEWNPNVK EIKVLQKIGK DTFITHELAA EAAGNLVGPR DFVSVRCAKR RGSTCVLAGM ATDFGNMPEQ KGVIRAEHGP TCMVLHPLAG SPSKTKLTWL LSIDLKGWLP KSIINQVLSQ TQVDFANHLR KRLESHPASE ARC | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The STAR solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 2mM DTT and 20% glycerol. |