missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SPRED1 Protein
A cDNA sequence encoding the SPRED1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13583-10ug
Additional Details : Weight : 0.01000kg
Specifications
SPRED1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSEETAT SDNDNSYARV RAVVMTRDDS SGGWLPLGGS GLSSVTVFKV PHQEENGCAD FFIRGERLRD KMVVLECMLK KDLIYNKVTP TFHHWKIDDK KFGLTFQSPA DARAFDRGIR RAIEDISQGC PESKNEAEGA DDLQANEEDS SSSLVKDHLF QQETVVTSEP YRSSNIRPSP FEDLNARRVY MQSQANQITF GQPGLDIQSR SMEYVQRQIS KECGSLKSQN RVPLKSIRHV SFQDEDEIVR INPRDILIRR YADYRHPDMW KNDLERDDAD SSIQFSKPDS KKSDYLYSCG DETKLSSPKD SVVFKTQPSS LKIKKSKRRK EDGERSRCVY CQERFNHEEN VRGKCQDAPD PIKRCIYQVS CMLCAESMLY HCMSDSEGDF SDPCSCDTSD DKFCLRWLAL VALSFIVPCM CCYVPLRMCH RCGEACGCCG GKHKAAG | |
Greater than 80.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SPRED1 solution (0.5 mg/1ml) contains 20mM Tris-HCl (pH 8.0) and 10% glycerol. |