missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SPRED1 Protein
A cDNA sequence encoding the SPRED1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Name | SPRED1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15974049
|
enQuireBio™
QP13583-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954049
|
enQuireBio™
QP13583-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964049
|
enQuireBio™
QP13583-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SPRED1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SPRED1 solution (0.5 mg/1ml) contains 20mM Tris-HCl (pH 8.0) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSEETAT SDNDNSYARV RAVVMTRDDS SGGWLPLGGS GLSSVTVFKV PHQEENGCAD FFIRGERLRD KMVVLECMLK KDLIYNKVTP TFHHWKIDDK KFGLTFQSPA DARAFDRGIR RAIEDISQGC PESKNEAEGA DDLQANEEDS SSSLVKDHLF QQETVVTSEP YRSSNIRPSP FEDLNARRVY MQSQANQITF GQPGLDIQSR SMEYVQRQIS KECGSLKSQN RVPLKSIRHV SFQDEDEIVR INPRDILIRR YADYRHPDMW KNDLERDDAD SSIQFSKPDS KKSDYLYSCG DETKLSSPKD SVVFKTQPSS LKIKKSKRRK EDGERSRCVY CQERFNHEEN VRGKCQDAPD PIKRCIYQVS CMLCAESMLY HCMSDSEGDF SDPCSCDTSD DKFCLRWLAL VALSFIVPCM CCYVPLRMCH RCGEACGCCG GKHKAAG | |
Greater than 80.0% as determined by SDS-PAGE. |