missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SPP1 Protein

A cDNA sequence encoding the SPP1 was constructed and used to recombinantly synthesize the protein.

£155.00 - £5730.00

Specifications

Name SPP1 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15954039
View Documents Promotion Details
enQuireBio™
QP13580-HIS-EC-10UG
10 μg
£155.00
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15964039
View Documents Promotion Details
enQuireBio™
QP13580-HIS-EC-1MG
1 mg
£5730.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

SPP1 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
Osteopontin was lyophilized from a 0.2microm filtered solution of 20mM PB and 150mM NaCl, pH 7.2.
Research Use Only
Recombinant Protein
E. coli
IPVKQADSGSSEEKQLYNKYPDAVATWLNPDPSQKQNLLAPQNAVSSEETNDFKQETLPSKSNESHDHMDDMDDEDDDDHVDSQDSIDSNDSDDVDDTDDSHQSDESHHSDESDELVTDFPTDLPATEVFTPVVPTVDTYDGRGDSVVYGLRSKSKKFRRPDIQYPDATDEDITSHMESEELNGAYKAIPVAQDLNAPSDWDSRGKDSYETSQLDDQSAETHSHKQSRLYKRKANDESNEHSDVIDSQELSKVSREFHSHEFHSHEDMLVVDPKSKEEDKHLKFRISHELDSASSEVNVDHHHHHH
Greater than 95.0% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications