missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Spike glycoProtein

A DNA sequence encoding the Homo sapiens (Human) Spike glycoprotein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7178-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16047580

  • £1527.00 / 1mg

Estimated Shipment: 25-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Spike glycoProtein
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Greater than 90% as determined by SDS-PAGE.
Research Use Only
S
Recombinant Protein
E. coli
VIGDLKCTSD NINDKDTGPP PISTDTVDVT NGLGTYYVLD RVYLNTTLFL NGYYPTSGST YRNMALKGSV LLSRLWFKPP FLSDFINGIF AKVKNTKVIK DRVMYSEFPA ITIGSTFVNT SYSVVVQPRT INSTQDGDNK LQGLLEVSVC QYNMCEYPQT ICHPNLGNHR KELWHLDTGV VSCLYKRNFT YDVNADYLYF HFYQEGGTFY AYFTDTGVVT KFLFNVYLGM ALSHYYVMPL TCNSKLTLEY WVTPLTSRQY LLAFNQDGII FNAEDCMSDF MSEIKCKTQS IAPPTGVYEL NGYTVQPIAD VYRRKPNLPN CNIEAWLNDK
Videos
SDS
Documents
Certificates