missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SP100 Protein
A cDNA sequence encoding the SP100 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13565-20ug
Additional Details : Weight : 0.01000kg
Specifications
SP100 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MAGGGGDLST RRLNECISPV ANEMNHLPAH SHDLQRMFTE DQGVDDRLLY DIVFKHFKRN KVEISNAIKK TFPFLEGLRD RDLITNKMFE DSQDSCRNLV PVQRVVYNVL SELEKTFNLP VLEALFSDVN MQEYPDLIHI YKGFENVIHD KLPLQESEEE EREERSGLQL SLEQGTGENS FRSLTWPPSG SPSHAGTTPP ENGLSEHPCE TEQINAKRKD TTSDKDDSLG SQQTNEQCAQ KAEPTESCEQ IAVQVNNGDA GREMPCPLPC DEESPEAELH NHGIQINSCS VRLVDIKKEK PFSNSKVECQ AQARTHHNQA SDIIVISSED SEGSTDVDEP LEVFISAPRS EPVINNDNPL ESNDEKEGQE ATCSRPQIVP EPMDFRKLST FRESFKKRVI GQDHDFSESS EEEAPAEASS GALRSKHGEK APMTSRSTST WRIPSRKRRF SSSDFSDLSN GEELQETCSS SLRRGSGSQP QEPENKKCSC VMCFPKGVPR SQEARTESSQ ASDMMDTMDV ENNSTLEKHS GKRRKKRRHR SKVNGLQRGR KKDRPRKHLT LNNKVQKKRW QQRGRKANTR PLKRRRKRGP RIPKDENINF KQSELPVTCG EVKGTLYKER FKQGTSKKCI QSEDKKWFTP REFEIEGDRG ASKNWKLSIR CGGYTLKVLM ENKFLPEPPS TRKKRILESH NNTLVDPCEE HKKKNPDASV KFSEFLKKCS ETWKTIFAKE KGKFEDMAKA DKAHYEREMK TYIPPKGEKK KKFKDPNAPK RPPLAFFLFC SEYRPKIKGE HPGLSIDDVV KKLAGMWNNT AAADKQFYEK KAAKLKEKYK KDIAAYRAKG KPNSAKKRVV KAEKSKKKKE EEEDEEDEQE EENEEDDDK | |
Greater than 80% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
Sp100 is supplied in 16mM HEPES buffer pH 8.0, 400mM NaCl, and 20% glycerol. |