missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SNCB Protein
A cDNA sequence encoding the SNCB was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13536-50ug
Additional Details : Weight : 0.01000kg
Specifications
SNCB Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MDVFMKGLSM AKEGVVAAAE KTKQGVTEAA EKTKEGVLYV GSKTREGVVQ GVASVAEKTK EQASHLGGAV FSGAGNIAAA TGLVKREEFP TDLKPEEVAQ EAAEEPLIEP LMEPEGESYE DPPQEEYQEY EPEA | |
Greater than 95.0% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The protein (1 mg/ml) contains 20mM Tris-HCl buffer (pH 7.5), 0.1M NaCl and 1mM MgCl2. |