missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SNAI2 Protein

A cDNA sequence encoding the SNAI2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13530-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15953869

  • £2978.71 / 1mg
Estimated Shipment: 12-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

SNAI2 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
SNAI2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl Buffer (pH 8.0) and 10% glycerol.
Greater than 80.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMPRSFLV KKHFNASKKP NYSELDTHTV IISPYLYESY SMPVIPQPEI LSSGAYSPIT VWTTAAPFHA QLPNGLSPLS GYSSSLGRVS PPPPSDTSSK DHSGSESPIS DEEERLQSKL SDPHAIEAEK FQCNLCNKTY STFSGLAKHK QLHCDAQSRK SFSCKYCDKE YVSLGALKMH IRTHTLPCVC KICGKAFSRP WLLQGHIRTH TGEKPFSCPH CNRAFADRSN LRAHLQTHSD VKKYQCKNCS KTFSRMSLLH KHEESGCCVA H
Videos
SDS
Documents
Certificates
Special Offers

Special Offers