missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Small muscular Protein
A DNA sequence encoding the Homo sapiens (Human) Small muscular protein, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_055147.1 |
---|---|
Gene ID (Entrez) | 23676 |
Name | Small muscular Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16003291
|
enQuireBio™
QP8209-EC-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16023291
|
enQuireBio™
QP8209-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16013291
|
enQuireBio™
QP8209-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033291
|
enQuireBio™
QP8209-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16043291
|
enQuireBio™
QP8209-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16053291
|
enQuireBio™
QP8209-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_055147.1 | |
Small muscular Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
23676 | |
Research Use Only | |
SMPX | |
Recombinant Protein | |
E. coli | |
MNMSKQPVSN VRAIQANINI PMGAFRPGAG QPPRRKECTP EVEEGVPPTS DEEKKPIPGA KKLPGPAVNL SEIQNIKSEL KYVPKAEQ | |
Greater than 80% as determined by SDS-PAGE. |