missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Small muscular Protein

A DNA sequence encoding the Homo sapiens (Human) Small muscular protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8209-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16023291

  • £2480.00 / 1mg
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_055147.1
Small muscular Protein
Research Use Only
SMPX
Recombinant Protein
E. coli
MNMSKQPVSN VRAIQANINI PMGAFRPGAG QPPRRKECTP EVEEGVPPTS DEEKKPIPGA KKLPGPAVNL SEIQNIKSEL KYVPKAEQ
Greater than 80% as determined by SDS-PAGE.
23676
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers