missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SLC25A20 Protein
A DNA sequence encoding the Homo sapiens (Human) SLC25A20, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6693-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_000378.1 | |
SLC25A20 Protein | |
Research Use Only | |
SLC25A20 | |
Recombinant Protein | |
E. coli | |
MADQPKPISP LKNLLAGGFG GVCLVFVGHP LDTVKVRLQT QPPSLPGQPP MYSGTFDCFR KTLFREGITG LYRGMAAPII GVTPMFAVCF FGFGLGKKLQ QKHPEDVLSY PQLFAAGMLS GVFTTGIMTP GERIKCLLQI QASSGESKYT GTLDCAKKLY QEFGIRGIYK GTVLTLMRDV PASGMYFMTY EWLKNIFTPE GKRVSELSAP RILVAGGIAG IFNWAVAIPP DVLKSRFQTA PPGKYPNGFR DVLRELIRDE GVTSLYKGFN AVMIRAFPAN AACFLGFEVA MKFLNWATPN L | |
Greater than 80% as determined by SDS-PAGE. |
788 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |