missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SLC25A20 Protein

A DNA sequence encoding the Homo sapiens (Human) SLC25A20, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6693-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005170

  • £321.05 / 50µg

Estimated Shipment
05-11-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

NP_000378.1
SLC25A20 Protein
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
788
Greater than 80% as determined by SDS-PAGE.
Research Use Only
SLC25A20
Recombinant Protein
E. coli
MADQPKPISP LKNLLAGGFG GVCLVFVGHP LDTVKVRLQT QPPSLPGQPP MYSGTFDCFR KTLFREGITG LYRGMAAPII GVTPMFAVCF FGFGLGKKLQ QKHPEDVLSY PQLFAAGMLS GVFTTGIMTP GERIKCLLQI QASSGESKYT GTLDCAKKLY QEFGIRGIYK GTVLTLMRDV PASGMYFMTY EWLKNIFTPE GKRVSELSAP RILVAGGIAG IFNWAVAIPP DVLKSRFQTA PPGKYPNGFR DVLRELIRDE GVTSLYKGFN AVMIRAFPAN AACFLGFEVA MKFLNWATPN L
Videos
Documents