missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SIRT1 / SIR2L1 Protein

A DNA sequence encoding the Homo sapiens (Human) SIRT1 / SIR2L1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP1351-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15953799

  • £1735.00 / 1mg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001135970.1
SIRT1 / SIR2L1 Protein
Research Use Only
SIRT1
Recombinant Protein
E. coli
ADEAALALQP GGSPSAAGAD REAASSPAGE PLRKRPRRDG PGLERSPGEP GGAAPEREVP AAARGCPGAA AAALWREAEA EAAAAGGEQE AQATAAAGEG DNGPGLQGPS REPPLADNLY DEDDDDEGEE EEEAAAAAIG YRDNLLFGDE IITNGFHSCE SDEEDRASHA SSSDWTPRPR IGPYTFVQQH LMIGTDPRTI LKDLLPETIP PPELDDMTLW QIVINILSEP PKRKKRKDIN TIEDAVKLLQ ECKKIIVLTG AGVSVSCGIP DFRSRDGIYA RLAVDFPDLP DPQAMFDIEY FRKDPRPFFK FAKEIYPGQF QPSLCHKFIA LSDKEGKLLR NYTQNIDTLE QVAGIQRIIQ CHGSFATASC LICKYKVDCE AVRGDIFNQV VPRCPRCPAD EPLAIMKPEI VFFGENLPEQ FHRAMKYDKD EVDLLIVIGS SLKVRPVALI PSSIPHEVPQ ILINREPLPH LHFDVELLGD CDVIINELCH RLGGEYAKLC CNPVKLSEIT EKPPRTQKEL AYLSELPPTP LHVSEDSSSP ERTSPPDSSV IVTLLDQAAK SNDDLDVSES KGCMEEKPQE VQTSRNVESI AEQMENPDLK NVGSSTGEKN ERTSVAGTVR KCWPNRVAKE QISRRLDGNQ YLFLPPNRYI FHGAEVYSDS EDDVLSSSSC GSNSDSGTCQ SPSLEEPMED ESEIEEFYNG LEDEPDVPER AGGAGFGTDG DDQEAINEAI SVKQEVTDMN YPSNKS
Greater than 90% as determined by SDS-PAGE.
23411
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers