missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human sIL 6R Protein

A cDNA sequence encoding the sIL 6R was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13504-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15963779

  • £155.00 / 20µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

3570
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MLAVGCALLA ALLAAPGAAL APRRCPAQEV ARGVLTSLPG DSVTLTCPGV EPEDNATVHW VLRKPAAGSH PSRWAGMGRR LLLRSVQLHD SGNYSCYRAG RPAGTVHLLV DVPPEEPQLS CFRKSPLSNV VCEWGPRSTP SLTTKAVLLV RKFQNSPAED FQEPCQYSQE SQKFSCQLAV PEGDSSFYIV SMCVASSVGS KFSKTQTFQG CGILQPDPPA NITVTAVARN PRWLSVTWQD PHSWNSSFYR LRFELRYRAE RSKTFTTWMV KDLQHHCVIH DAWSGLRHVV QLRAQEEFGQ GEWSEWSPEA MGTPWTESRS PPAENEVSTP MQALTTNKDD DNILFRDSAN ATSLPVQDSS SVPLPTFLVA GGSLAFGTLL CIAIVLRFKK TWKLRALKEG KTSMHPPYSL GQLVPERPRP TPVLVPLISP PVSPSSLGSD NTSSHNRPDA RDPRSPYDIS NTDYFFPR
Greater than 90.0% as determined by SDS-PAGE.
sIL 6R Protein
Research Use Only
IL6R
Human
His
CD126 His-Tag protein is supplied in 20mM Tris-HCl, pH 8.0, 0.8M Urea and 50% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers