missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human sIL 6R Protein
A cDNA sequence encoding the sIL 6R was constructed and used to recombinantly synthesize the protein.
£99.15 - £3085.00
Specifications
Gene ID (Entrez) | 3570 |
---|---|
Name | sIL 6R Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | IL6R |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15973779
|
enQuireBio™
QP13504-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15963779
|
enQuireBio™
QP13504-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15953779
|
enQuireBio™
QP13504-1MG |
1 mg |
£3085.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
3570 | |
Research Use Only | |
IL6R | |
Human | |
His | |
CD126 His-Tag protein is supplied in 20mM Tris-HCl, pH 8.0, 0.8M Urea and 50% glycerol. |
sIL 6R Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MLAVGCALLA ALLAAPGAAL APRRCPAQEV ARGVLTSLPG DSVTLTCPGV EPEDNATVHW VLRKPAAGSH PSRWAGMGRR LLLRSVQLHD SGNYSCYRAG RPAGTVHLLV DVPPEEPQLS CFRKSPLSNV VCEWGPRSTP SLTTKAVLLV RKFQNSPAED FQEPCQYSQE SQKFSCQLAV PEGDSSFYIV SMCVASSVGS KFSKTQTFQG CGILQPDPPA NITVTAVARN PRWLSVTWQD PHSWNSSFYR LRFELRYRAE RSKTFTTWMV KDLQHHCVIH DAWSGLRHVV QLRAQEEFGQ GEWSEWSPEA MGTPWTESRS PPAENEVSTP MQALTTNKDD DNILFRDSAN ATSLPVQDSS SVPLPTFLVA GGSLAFGTLL CIAIVLRFKK TWKLRALKEG KTSMHPPYSL GQLVPERPRP TPVLVPLISP PVSPSSLGSD NTSSHNRPDA RDPRSPYDIS NTDYFFPR | |
Greater than 90.0% as determined by SDS-PAGE. |