missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Signal-regulatory protein delta Protein
A DNA sequence encoding the Homo sapiens (Human) Signal-regulatory protein delta, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6686-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_848555.2 | |
Signal-regulatory protein delta Protein | |
Research Use Only | |
SIRPD | |
Recombinant Protein | |
E. coli | |
FHVQQTEMSQ TVSTGESIIL SCSVPNTLPN GPVLWFKGTG PNRKLIYNFK QGNFPRVKEI GDTTKPGNTD FSTRIREISL ADAGTYYCVK FIKGRAIKEY QSGRGTQVFV TEQNPRPPKN RPAGRAGSRA HHDAHTCLSA LPERNSTNYF VQPCCCLRLL GLTGLLSK | |
Greater than 90% as determined by SDS-PAGE. |
128646 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |