missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Signal-regulatory protein delta Protein

A DNA sequence encoding the Homo sapiens (Human) Signal-regulatory protein delta, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6686-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16085100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_848555.2
Signal-regulatory protein delta Protein
Research Use Only
SIRPD
Recombinant Protein
E. coli
FHVQQTEMSQ TVSTGESIIL SCSVPNTLPN GPVLWFKGTG PNRKLIYNFK QGNFPRVKEI GDTTKPGNTD FSTRIREISL ADAGTYYCVK FIKGRAIKEY QSGRGTQVFV TEQNPRPPKN RPAGRAGSRA HHDAHTCLSA LPERNSTNYF VQPCCCLRLL GLTGLLSK
Greater than 90% as determined by SDS-PAGE.
128646
200 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers