Learn More
enQuireBio™ Recombinant Human SHH / Sonic hedgehog Protein
Recombinant Protein
Brand:  enQuireBio™ QP5292-1mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15945859
  • £2865.00 / 1mg
Estimated Shipment: 05-06-2024
to see stock.
Specifications
NP_000184.1
SHH / Sonic hedgehog Protein
Research Use Only
SHH
Recombinant Protein
E. coli
MIIGPGRGFG KRRHPKKLTP LAYKQFIPNV AEKTLGASGR YEGKISRNSE RFKELTPNYN PDIIFKDEEN TGADRLMTQR CKDKLNALAI SVMNQWPGVK LRVTEGWDED GHHSEESLHY EGRALDITTS DRDRSKYGML ARLAVEAGFD WVYYESKAHI HCSVKAENSV AAKSGG
Greater than 95% as determined by SDS-PAGE.
6469
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity at a concentration of 5 ug/mL in the CCL-226 cell proliferation assay. The bioactivity acceptance criteria is set at 200 units/mg.
Human
Untagged
10 mM sodium phosphate, pH 7.5