missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SH3BGRL Protein

A cDNA sequence encoding the SH3BGRL was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13487-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15953729

  • £155.00 / 20µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

SH3BGRL Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMVIRVY IASSSGSTAI KKKQQDVLGF LEANKIGFEE KDIAANEENR KWMRENVPEN SRPATGYPLP PQIFNESQYR GDYDAFFEAR ENNAVYAFLG LTAPPGSKEA EVQAKQQA
Greater than 95.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
SH3BGRL protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.15M NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers