missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SGTA Protein
A cDNA sequence encoding the SGTA was constructed and used to recombinantly synthesize the protein.
£99.15 - £2490.00
Specifications
Name | SGTA Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15973719
|
enQuireBio™
QP13484-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15963719
|
enQuireBio™
QP13484-25UG |
25 μg |
£155.00
25µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15953719
|
enQuireBio™
QP13484-1MG |
1 mg |
£2490.00
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SGTA Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SGTA Human solution containing 20mM Trsi HCL pH 8.0, 1mM DTT and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MDNKKRLAYA IIQFLHDQLR HGGLSSDAQE SLEVAIQCLE TAFGVTVEDS DLALPQTLPE IFEAAATGKE MPQDLRSPAR TPPSEEDSAE AERLKTEGNE QMKVENFEAA VHFYGKAIEL NPANAVYFCN RAAAYSKLGN YAGAVQDCER AICIDPAYSK AYGRMGLALS SLNKHVEAVA YYKKALELDP DNETYKSNLK IAELKLREAP SPTGGVGSFD IAGLLNNPGF MSMASNLMNN PQIQQLMSGM ISGGNNPLGT PGTSPSQNDL ASLIQAGQQF AQQMQQQNPE LIEQLRSQIR SRTPSASNDD QQELEHHHHH H | |
Greater than 95% as determined by SDS-PAGE. |