missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Serum paraoxonase / arylesterase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Serum paraoxonase/arylesterase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6513-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000437.3 | |
Serum paraoxonase / arylesterase 1 Protein | |
Research Use Only | |
PON1 | |
Recombinant Protein | |
E. coli | |
AKLIALTLLG MGLALFRNHQ SSYQTRLNAL REVQPVELPN CNLVKGIETG SEDLEILPNG LAFISSGLKY PGIKSFNPNS PGKILLMDLN EEDPTVLELG ITGSKFDVSS FNPHGISTFT DEDNAMYLLV VNHPDAKSTV ELFKFQEEEK SLLHLKTIRH KLLPNLNDIV AVGPEHFYGT NDHYFLDPYL QSWEMYLGLA WSYVVYYSPS EVRVVAEGFD FANGINISPD GKYVYIAELL AHKIHVYEKH ANWTLTPLKS LDFNTLVDNI SVDPETGDLW VGCHPNGMKI FFYDSENPPA SEVLRIQNIL TEEPKVTQVY AENGTVLQGS TVASVYKGKL LIGTVFHKAL YCEL | |
Greater than 90% as determined by SDS-PAGE. |
5444 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |