missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SerpinB4 Protein

A DNA sequence encoding the Homo sapiens (Human) SerpinB4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6670-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16004980

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_002965.1
SerpinB4 Protein
Research Use Only
SERPINB4
Recombinant Protein
E. coli
MNSLSEANTK FMFDLFQQFR KSKENNIFYS PISITSALGM VLLGAKDNTA QQISKVLHFD QVTENTTEKA ATYHVDRSGN VHHQFQKLLT EFNKSTDAYE LKIANKLFGE KTYQFLQEYL DAIKKFYQTS VESTDFANAP EESRKKINSW VESQTNEKIK NLFPDGTIGN DTTLVLVNAI YFKGQWENKF KKENTKEEKF WPNKNTYKSV QMMRQYNSFN FALLEDVQAK VLEIPYKGKD LSMIVLLPNE IDGLQKLEEK LTAEKLMEWT SLQNMRETCV DLHLPRFKME ESYDLKDTLR TMGMVNIFNG DADLSGMTWS HGLSVSKVLH KAFVEVTEEG VEAAAATAVV VVELSSPSTN EEFCCNHPFL FFIRQNKTNS ILFYGRFSSP
Greater than 90% as determined by SDS-PAGE.
6318
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers