missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SERPINA4 Protein
A cDNA sequence encoding the SERPINA4 was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Name | SERPINA4 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15963629
|
enQuireBio™
QP13457-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15943629
|
enQuireBio™
QP13457-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15953629
|
enQuireBio™
QP13457-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SERPINA4 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SERPINA4 was filtered (0.4microm) and lyophilized from 0.5 mg/ml solution in phosphate buffered saline pH 7.4 and 5% (w/v) Trehalose. |
Research Use Only | |
Recombinant Protein | |
Human (HEK293) | |
QLHVEHDGES CSNSSHQQIL ETGEGSPSLK IAPANADFAF RFYYLIASET PGKNIFFSPL SISAAYAMLS LGACSHSRSQ ILEGLGFNLT ELSESDVHRG FQHLLHTLNL PGHGLETRVG SALFLSHNLK FLAKFLNDTM AVYEAKLFHT NFYDTVGTIQ LINDHVKKET RGKIVDLVSE LKKDVLMVLV NYIYFKALWE KPFISSRTTP KDFYVDENTT VRVPMMLQDQ EHHWYLHDRY LPCSVLRMDY KGDATVFFIL PNQGKMREIE EVLTPEMLMR WNNLLRKRNF YKKLELHLPK FSISGSYVLD QILPRLGFTD LFSKWADLSG ITKQQKLEAS KSFHKATLDV DEAGTEAAAA TSFAIKFFSA QTNRHILRFN RPFLVVIFST STQSVLFLGK VVDPTKPHHH HHHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |