missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SERPINA4 Protein
A cDNA sequence encoding the SERPINA4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13457-1mg
Additional Details : Weight : 0.01000kg
Specifications
SERPINA4 Protein | |
Research Use Only | |
Recombinant Protein | |
Human (HEK293) | |
QLHVEHDGES CSNSSHQQIL ETGEGSPSLK IAPANADFAF RFYYLIASET PGKNIFFSPL SISAAYAMLS LGACSHSRSQ ILEGLGFNLT ELSESDVHRG FQHLLHTLNL PGHGLETRVG SALFLSHNLK FLAKFLNDTM AVYEAKLFHT NFYDTVGTIQ LINDHVKKET RGKIVDLVSE LKKDVLMVLV NYIYFKALWE KPFISSRTTP KDFYVDENTT VRVPMMLQDQ EHHWYLHDRY LPCSVLRMDY KGDATVFFIL PNQGKMREIE EVLTPEMLMR WNNLLRKRNF YKKLELHLPK FSISGSYVLD QILPRLGFTD LFSKWADLSG ITKQQKLEAS KSFHKATLDV DEAGTEAAAA TSFAIKFFSA QTNRHILRFN RPFLVVIFST STQSVLFLGK VVDPTKPHHH HHHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SERPINA4 was filtered (0.4microm) and lyophilized from 0.5 mg/ml solution in phosphate buffered saline pH 7.4 and 5% (w/v) Trehalose. |